Anti-TP53I11 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A43950-100UL A43950-50UL
77706-803EA 334.95 USD
77706-803 77706-804
Anti-TP53I11 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TP53I11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR