Gold ≥99.99%, disk, as rolled, Diameter 35 mm

Supplier: GOODFELLOW CORPORATION

1000046575 1000047538
77681-691EA 2828.31 USD
77681-691 77681-913
Gold ≥99.99%, disk, as rolled, Diameter 35 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR