Gold ≥99.99%, disk, as rolled, Diameter 20.7 mm

Supplier: GOODFELLOW CORPORATION

1000047479 1000046526
77681-890EA 1271.25 USD
77681-890 77681-667
Gold ≥99.99%, disk, as rolled, Diameter 20.7 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR