Gold ≥99.99%, disk, as rolled, Diameter 15 mm

Supplier: GOODFELLOW CORPORATION

1000030556 1000019909 1000001128 1000003772 1000032072 1000030657
77680-817EA 304.49 USD
77680-817 77680-719 77680-691 77680-696 77681-170 77680-846
Gold ≥99.99%, disk, as rolled, Diameter 15 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR