Gold ≥99.99%, disk, as rolled, Diameter 12.7 mm

Supplier: GOODFELLOW CORPORATION

1000030554 1000030688 1000031557 1000032070 1000030656 1000031909
77680-815EA 206.59 USD
77680-815 77680-859 77681-039 77681-169 77680-845 77681-128
Gold ≥99.99%, disk, as rolled, Diameter 12.7 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR