Gold ≥99.95%, disk, as rolled, Diameter 9.2 mm
Supplier: GOODFELLOW CORPORATION
1000043997
1000043513
1000046681
1000046892
77681-354EA
331.46
USD
77681-354
77681-278
77681-715
77681-789
Gold ≥99.95%, disk, as rolled, Diameter 9.2 mm
Gold
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 Melting Pt: 1063 Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...