Gold ≥99.95%, disk, as rolled, Diameter 28 mm

Supplier: GOODFELLOW CORPORATION

1000018897 1000043585 1000046802 1000047007
77680-715EA 1399.94 USD
77680-715 77681-315 77681-753 77681-827
Gold ≥99.95%, disk, as rolled, Diameter 28 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR