Gold ≥99.999%, disk, as rolled, Diameter 15.5 mm

Supplier: GOODFELLOW CORPORATION

1000041423 1000044908 1000044211 1000046296
77681-234EA 3371.43 USD
77681-234 77681-513 77681-441 77681-583
Gold ≥99.999%, disk, as rolled, Diameter 15.5 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR