Gold ≥99.999%, disk, as rolled, Diameter 9.2 mm
Supplier: GOODFELLOW CORPORATION
1000046263
1000041383
1000044879
1000044180
77681-570EA
348.87
USD
77681-570
77681-219
77681-499
77681-427
Gold ≥99.999%, disk, as rolled, Diameter 9.2 mm
Gold
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 Melting Pt: 1063 Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...