Gold ≥99.9%, disk, Diameter 8 mm

Supplier: GOODFELLOW CORPORATION

1000030325 1000030428 1000030382 1000030946 1000030985 1000004531 1000014798 1000031032 1000031341 1000031839 1000031189 1000031755 1000030832 1000031652 1000031798
77680-746EA 153.33 USD
77680-746 77680-787 77680-772 77680-921 77680-929 77680-697 77680-705 77680-938 77681-012 77681-110 77680-977 77681-084 77680-899 77681-065 77681-095
Gold ≥99.9%, disk, Diameter 8 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR