Gold ≥99.9%, disk, Diameter 6 mm
Supplier: GOODFELLOW CORPORATION
1000030324
1000030426
1000030380
1000030531
1000030943
1000030982
1000031029
1000031338
1000031186
1000031753
1000030474
1000031837
1000030829
1000031649
1000031795
77680-745EA
139.73
USD
77680-745
77680-786
77680-771
77680-812
77680-920
77680-928
77680-937
77681-011
77680-976
77681-083
77680-799
77681-109
77680-898
77681-064
77681-094
Gold ≥99.9%, disk, Diameter 6 mm
Gold
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 Melting Pt: 1063 Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...