Gold ≥99.9%, disk, Diameter 6.35 mm

Supplier: GOODFELLOW CORPORATION

1000030323 1000030425 1000030378 1000030530 1000030979 1000031026 1000031335 1000030940 1000031183 1000031751 1000030473 1000031836 1000030826 1000031646 1000031792
77680-744EA 141.97 USD
77680-744 77680-785 77680-770 77680-811 77680-927 77680-936 77681-010 77680-919 77680-975 77681-082 77680-798 77681-108 77680-897 77681-063 77681-093
Gold ≥99.9%, disk, Diameter 6.35 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR