Gold ≥99.9%, disk, Diameter 10 mm

Supplier: GOODFELLOW CORPORATION

1000030454 1000030401 1000017982 1000030358 1000030964 1000030511 1000031004 1000030921 1000031168 1000030302 1000030791 1000031611 1000031729 1000031320 1000031816
77680-788EA 237.31 USD
77680-788 77680-773 77680-713 77680-758 77680-922 77680-800 77680-930 77680-914 77680-970 77680-732 77680-885 77681-052 77681-072 77681-005 77681-096
Gold ≥99.9%, disk, Diameter 10 mm
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR