Blood Pressure Sensors
Supplier: WORLD PRECISION INSTRUMENTS LLC
II-B60-025 II-B60-038 II-B60-044 II-B63-075
77664-436EA 550 USD
77664-436 77664-438 77664-440 77664-442
Blood Pressure Sensors
Blood Pressure Monitors
The tail cuff sensor has the ability to detect mouse/rat tail cuff blood pressure via the tail photoelectrically.
  • All the sensors are nonmagnetic
  • They are extremely sensitive in detecting pulses with the use of the II-MRBP systems
  • The ambient temperature that a reading can be taken starts at 30 °C
The systolic readings are within 5 mm Hg of cannulation and telemetry (Validated).