6-Amino-2H-1,4-benzoxazin-3(4H)-one ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 6-Amino-3,4-dihydro-3-oxo-2H-1,4-benzoxazine,6-Amino-3-oxo-3,4-dihydrobenzo[1,4]oxazine,6-Amino-2H-benzo[b][1,4]oxazin-3(4H)-one,6-Amino-4H-benzo[1,4]oxazin-3-one, 6-amino-2,4-dihydro-1,4-benzoxazin-3-one

A187938-10G A187938-1G A187938-25G A187938-5G
ALA187938-10GEA 523.36 USD
ALA187938-10G ALA187938-1G ALA187938-25G ALA187938-5G
6-Amino-2H-1,4-benzoxazin-3(4H)-one ≥97%
6-Amino-2H-1,4-benzoxazin-3(4H)-one
Formula: C₈H₈N₂O₂
MW: 164.17 g/mol
Melting Pt: 88-93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425742
CAS Number: 89976-75-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=432f20468850c8173ca60a085f37671078fad46e&vid=432f20468850c8173ca60a085f37671078fad46e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR