1-Acetyl-5-nitroindoline ≥98%

Supplier: ALADDIN SCIENTIFIC

A169675-5G A169675-1G
ALA169675-5GEA 127.21 USD
ALA169675-5G ALA169675-1G
1-Acetyl-5-nitroindoline ≥98%
1-Acetyl-5-nitroindoline
Formula: C₁₀H₁₀N₂O₃
Melting Pt: 175-178°C
Storage Temperature: Ambient
MDL Number: MFCD00005811
CAS Number: 33632-27-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c52f8ebf8f4acb66d268b5cedfd1cc8775510f16&vid=c52f8ebf8f4acb66d268b5cedfd1cc8775510f16&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR