3-Acetylcoumarin ≥98%
Supplier: ALADDIN SCIENTIFIC
Warning
|
FTIR and FT-Raman spectra of 3-acetylcoumarin has been reported. 3-Acetylcoumarin undergoes condensation with aryl aldehydes in chloroform in the presence of piperidine to yield coumarin derivatives containing 4-arylbut-3-en-2-one moiety.
Formula:
C₁₁H₈O₃ Melting Pt: 123 °C |
MDL Number:
MFCD00006853 CAS Number: 3949-36-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...