5-Amino-3-methylisoxazole ≥97% (by titration)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine

A151656-5G A151656-1G
ALA151656-5GEA 93.01 USD
ALA151656-5G ALA151656-1G
5-Amino-3-methylisoxazole ≥97% (by titration)
5-Amino-3-methylisoxazole

5-Amino-3-methylisoxazole reacts with α,β-unsaturated ketones to yield the corresponding isoxazolo[5,4-b]pyridines.

Formula: C₄H₆N₂O
MW: 98.1 g/mol
Boiling Pt: 194
Melting Pt: 84
MDL Number: MFCD00003151
CAS Number: 14678-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eaba2b58ca2c33be9fa4611f39e621d1777c980f&vid=eaba2b58ca2c33be9fa4611f39e621d1777c980f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR