2-Amino-1-methoxybutane ≥90% (by GC)
Supplier: Aladdin Scientific
|
Danger
|
A151357-100ML
A151357-25ML
A151357-5ML
ALA151357100MLEA
113.91
USD
ALA151357100ML
ALA151357-25ML
ALA151357-5ML
2-Amino-1-methoxybutane ≥90% (by GC)
2-Amino-1-methoxybutane
Formula:
C₅H₁₃NO Density: 0.89 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00047920 CAS Number: 63448-63-5 UN: 2734 ADR: 8(3),II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...