2-Acetyl-3,5(6)-dimethylpyrazine ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A135006-1G
A135006-25G
A135006-5G
ALA135006-1GEA
43.61
USD
ALA135006-1G
ALA135006-25G
ALA135006-5G
2-Acetyl-3,5(6)-dimethylpyrazine ≥98%
2-Acetyl-3,5(6)-dimethylpyrazine
Formula:
C₁₆H₂₀N₄O₂ Boiling Pt: 70 °C Density: 1.08 g/cm³ Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD29036871 CAS Number: 72797-17-2 EINECS: 259-076-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...