5-Acetyluracil ≥97%
Supplier: ALADDIN SCIENTIFIC
Synonyms:
5-Acetylpyrimidine-2,4(1H,3H)-dione
A134088-1G
A134088-25G
A134088-5G
ALA134088-1GEA
47.41
USD
ALA134088-1G
ALA134088-25G
ALA134088-5G
5-Acetyluracil ≥97%
5-Acetyluracil
Formula:
C₆H₆N₂O₃ Melting Pt: 278°C |
MDL Number:
MFCD00151968 CAS Number: 6214-65-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...