Adefovir dipivoxil ≥99%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Adefovir Dipivoxil, 9-(2[bis(Pivaloyloxymethoxy)phosphorylmethoxy]ethyl)adenine, 9-[2-[Bis(pivaloyloxymethoxy)phosphorylmethoxy]ethyl]adenine, ((((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphoryl)bis(oxy))bis(methylene) bis(2,2-dimethylpropanoate)
Adefovir Dipivoxil works by blocking reverse transcriptase, an enzyme that is crucial for the hepatitis B virus (HBV) to reproduce in the body. Target: NRTIs; HBV Adefovir Dipivoxil works by blocking reverse transcriptase, an enzyme that is crucial for the hepatitis B virus (HBV) to reproduce in the body. Adefovir Dipivoxil is used for treatment of hepatitis B and herpes simplex virus infection [1-3]. Adefovir Dipivoxil is approved for the treatment of chronic hepatitis B in adults with evidence of active viral replication and either evidence of persistent elevations in serum aminotransferases (primarily ALT) or histologically active disease. Adefovir Dipivoxil is a failed treatment for HIV[3, 4]. Catalyst employed in the Sharpless Asymmetric Dihyroxylation of (E,E)- or (E,Z)-1,3-dienoates.1 Also used in the preparation of ?-hydroxy-γ-lactones from ?,γ-unsaturated esters.2Product Introduction:Adefovir dipivoxil, an adenosine analogue, is an oral proagent of the nucleoside reverse transcriptase inhibitor Adefovir. Adefovir dipivoxil inhibits both the wild type and HBV Lamivudine-resistant strains. Adefovir dipivoxil shows anti-orthopoxvirus activity.
Formula:
C₂₀H₃₂N₅O₈P MW: 501.48 g/mol Melting Pt: 101 °C Storage Temperature: Freezer |
MDL Number:
MFCD00869897 CAS Number: 142340-99-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...