AZD-8055 ≥99%, Moligand™
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
(5-(2,4-Bis((S)-3-methylmorpholino)pyrido[2,3-d]pyrimidin-7-yl)-2-methoxyphenyl)methanol, AZD 8055, (5-(2,4-bis((3s)-3-methylmorpholin-4-yl)pyrido(2,3-d)pyrimidin-7-yl)-2-methoxyphenyl)methanol, AZD8055, CCG168
A126366-200MG
A126366-50MG
ALA126366200MGEA
409.36
USD
ALA126366200MG
ALA126366-50MG
AZD-8055 ≥99%, Moligand™
AZD-8055
AZD8055 is a novel ATP-competitive mTOR inhibitor with IC50 of 0.8 nM with excellent selectivity (~1,000-fold) against PI3K isoforms and ATM/DNA-PK.
Formula:
C₂₅H₃₁N₅O₄ Density: 1.248 g/cm³ Storage Temperature: Freezer |
MDL Number:
MFCD16660191 CAS Number: 1009298-09-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...