Antazoline hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A124055-1G A124055-25G A124055-5G
ALA124055-1GEA 9.41 USD
ALA124055-1G ALA124055-25G ALA124055-5G
Antazoline hydrochloride ≥98%
Antazoline hydrochloride

Imidazoline agonist; more potent than efaroxan in inducing insulin release from β cells; H1 histamine receptor antagonist.Imidazoline binding site ligand and an antihistamine

Formula: C₁₇H₁₉N₃
Melting Pt: 238°C
Storage Temperature: Freezer
MDL Number: MFCD00058145
CAS Number: 2508-72-7
EINECS: 219-719-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=807099af2da58f30c89c392c6c641dd306daac6e&vid=807099af2da58f30c89c392c6c641dd306daac6e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR