2-Amino-5-bromonicotinonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

A123164-1G A123164-250MG A123164-5G
ALA123164-1GEA 386.56 USD
ALA123164-1G ALA123164250MG ALA123164-5G
2-Amino-5-bromonicotinonitrile ≥97%
2-Amino-5-bromonicotinonitrile
Formula: C₆H₄BrN₃
MDL Number: MFCD08688591
CAS Number: 709652-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bd671a8b5e985e45386f460efa6d7008900746c&vid=8bd671a8b5e985e45386f460efa6d7008900746c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR