2-Aminonicotinonitrile ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-Aminopyridine-3-carbonitrile, 2-Amino-3-cyanopyridine

A123148-1G A123148-5G
ALA123148-1GEA 19.86 USD
ALA123148-1G ALA123148-5G
2-Aminonicotinonitrile ≥98% (by GC)
2-Aminonicotinonitrile
Formula: C₆H₅N₃
MW: 119.13 g/mol
Melting Pt: 135°C
MDL Number: MFCD00128867
CAS Number: 24517-64-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0470969023ba22667b9fa99cc00c31e91d95c1be&vid=0470969023ba22667b9fa99cc00c31e91d95c1be&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR