4-Amino-3-(trifluoromethoxy)benzoic acid ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A120781-100G
A120781-10G
A120781-1G
A120781-25G
A120781-5G
ALA120781-100GEA
962.26
USD
ALA120781-100G
ALA120781-10G
ALA120781-1G
ALA120781-25G
ALA120781-5G
4-Amino-3-(trifluoromethoxy)benzoic acid ≥98%
4-Amino-3-(trifluoromethoxy)benzoic acid
Formula:
C₈H₆F₃NO₃ |
MDL Number:
MFCD00205143 CAS Number: 175278-22-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...