Kelly Hemostatic Forceps
Supplier: WORLD PRECISION INSTRUMENTS LLC
501714 501714-G 501715 501715-G 501288 501288-G 501241 501241-G
77662-698EA 58.52 USD
77662-698 77662-700 77662-702 77662-704 77662-560 77662-562 77662-472 77662-474
Kelly Hemostatic Forceps
Forceps
Kelly hemostatic forceps are used to clamp blood vessels and hold heavy tissues in place.
  • Serrated tips
  • Stainless steel
Kelly forceps, both straight and curved, are available. These forceps feature a locking ratchet and wider finger rings for easy handling.