Coverslips

Supplier: SARSTEDT INC
83.1840 83.1840.001 83.1840.002
77589-232CS 1184.09 USD
77589-232 77589-234 77589-236
Coverslips
Cover Glasses
To keep cells happy, the selection of a suitable cultivation surface is essential. SARSTEDT offers a wide range of cell culture labware and accessories for many applications.

  • Cell type: Adherent cells
  • Material: PET-G
  • Purity: Sterile, non-pyrogenic/endotoxin-free, non-cytotoxic
  • Made of unbreakable plastic for easy handling
  • Surface treatment on both sides (TC treatment) for better cell adhesion
  • Good optical properties
  • For use in cell culture dishes and plates

Coverslips are useful for cell cultivation, fixation and staining for microscopy.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR