2-Chloro-1-(2,6-difluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

471476-5G 471476-25G 471476-1G 471476-500MG
77633-278EA 716 USD
77633-278 77655-992 77643-098 77651-070
2-Chloro-1-(2,6-difluorophenyl)ethanone ≥95%
2-Chloro-1-(2,6-difluorophenyl)ethanone

Caution: For Research Use Only

Formula: C₈H₅ClF₂O
Storage Temperature: Ambient
CAS Number: 929249-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR