1-(3-Chloro-2,6-difluorophenyl)-2-methyl-1-propanone ≥95%
Supplier: AOBChem USA
|
Warning
|
460094-5G
460094-1G
460094-500MG
77629-496EA
1484.23
USD
77629-496
77642-958
77650-938
1-(3-Chloro-2,6-difluorophenyl)-2-methyl-1-propanone ≥95%
1-(3-Chloro-2,6-difluorophenyl)-2-methyl-1-propanone
Caution: For Research Use Only
Formula:
C₁₀H₉ClF₂O Storage Temperature: Ambient |
CAS Number:
2352647-02-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...