SwifTest™ DPD Chlorine Testing Systems
Supplier: Hach
2105560 2105660 2802400 2802300
104085-770PK 115.92 USD
104085-770 104085-774 104086-678 104088-308
SwifTest™ DPD Chlorine Testing Systems
Environmental Test Kits
Fast, easy reagent addition - ideal for high volume testing.
  • Just squeeze to add powder, no need to open reagent containers
Delivery information: Dispenser is supplied with 1 refill vial with enough reagent for approximately 250 tests.