Visual Comparator Test Kits, Chlorine
Supplier: Hach
1454200 2129000 223101 223102 223103
104085-492EA 242.14 USD
104085-492 104085-862 104085-952 104085-954 104085-956
Visual Comparator Test Kits, Chlorine
Environmental Test Kits
Qualitative results using traditional colour comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case for 100 tests
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Delivery information: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.