Nitrogen-Nitrate Standard Solution
Supplier: Hach
2415132 204649 1426010 194749
104086-208EA 29.48 USD
104086-208 104085-716 104085-404 104085-674
Nitrogen-Nitrate Standard Solution
Standards Environmental Analysis Standards
Standard solutions for accuracy checks in nitrate nitrogen determinations and also for meter calibrations/standardisation.