Reaction Tubes
Supplier: SARSTEDT INC
72.691.006 72.691 72.691.001 72.691.002 72.691.004 72.691.005 72.691.007
77588-762CS 356.17 USD
77588-762 77588-752 77588-754 77588-756 77588-758 77588-760 77588-764
Reaction Tubes
Tubes PCR Tubes
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Polypropylene retaining loop caps attached
  • Polypropylene tube material
  • Easy to open, secure seal when closed
  • Larger packaging units for economical storage and stocking
  • High centrifugation up to 20,000 ×g
This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base, with or without skirted base, with push cap or screw cap, with or without print or labels.