Screw Cap Tubes with Flat Base, PS
Supplier: SARSTEDT INC
60.9922.221 60.9921.532 60.9922.113 60.9922.115
77587-844CS 237.77 USD
77587-844 77587-830 77587-838 77587-840
Screw Cap Tubes with Flat Base, PS
Tubes Centrifuge Tubes
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Without graduations
  • Flat base shape ensures the tube stands securely
  • Polystyrene (PS) tubes for maximum transparency
  • O-ring cap increases security for fluctuations in temperature
This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base, with or without skirted base, with stopper or screw cap, with or without print or labels.