Screw Cap Tubes with Flat Base, PC

Supplier: SARSTEDT INC
60.9922.212 71.9923.610
77587-842CS 278.55 USD
77587-842 77588-592
Screw Cap Tubes with Flat Base, PC
Tubes Centrifuge Tubes
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.

  • Without graduations
  • Closures enclosed
  • Flat base shape
  • High temperature resistance up to 138 °C
  • Tubes and cap are autoclavable
  • Suitable for deep-freeze storage down to –80 °C
  • PP screw cap

This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR