Dissecting Scissors
Supplier: WORLD PRECISION INSTRUMENTS LLC
15922-G 15923 15923-G 15922 14393 14393-G 14394 14394-G
77661-956EA 53 USD
77661-956 77661-958 77661-960 77661-954 77661-886 77661-888 77661-890 77661-892
Dissecting Scissors
Scissors
Ring-handled dissecting scissors are surgical scissors commonly used to cut and separate delicate tissues. These scissors are 10 or 12.5 cm long. These dissecting scissors are available with straight or curved blades. These dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. Dissecting scissors have thinner (and often longer) blades than operating scissors.
  • Multi-purpose use
  • Ideal for cutting soft tissue or cartilage
  • Straight or curved
  • Long thin blades make them lighter than operating scissors
  • Surgical grade stainless steel or premium German steel
They are designed for cutting and probing in tight spaces. Their tips are pointed, where operating scissors are generally blunt. These are available in surgical grade stainless steel or premium German steel. Our Dissecting Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.