Nucleic Acid Purification Systems, Promega®
Supplier: Promega Corporation
A9281 A9282
PAA9281EA 159.46 USD
PAA9281 PAA9282
Nucleic Acid Purification Systems, Promega®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Silica Membrane-based Wizard® Plus SV Minipreps System isolates plasmid DNA.
  • Maxipreps and Megapreps kits Include Reservoirs Attached to Columns, 5 mL Syringes, and 0.22 µm Syringe Filters
  • Minipreps Kits come with 3 mL Syringes
  • MagneSil Paramagnetic Particles (PMPs) Enhance Binding Kinetics and Efficiency
  • SV Formats Work with a Spin or Vacuum Protocol
  • Wizard DNA Clean-Up System: Alternative to Phenol-Chloroform Extractions and Ethanol Precipitations
Wizard SV Gel and PCR Clean-Up System extracts and purifies DNA
Purified DNA works in radioactive sequencing, labeling, and cloning; enzyme reactions; DNA amplification and membrane hybridization; and PCR-based tests for Genetically Modified Organism (GMO) DNA sequences.
Wizard PCR Preps DNA Purification System purifies PCR items and DNA. Wizard DNA Clean-Up System purifies DNA from restriction digests and more and contains DNA resin, minicolumns, and syringe barrels. Wizard Genomic DNA Purification Kit features cell and nucleic lysis, protein precipitation, DNA rehydration, and solution to purify DNA. Wizard SV Genomic DNA Purification System features solution, buffer, concentrated wash and RNase A solution, and nuclease-free water. Wizard Magnetic 96 DNA Plant System purifies DNA from leaf/seed tissue and features MagneSil® Paramagnetic Particles (PMPs), lysis buffer A and B, and wash buffer. Wizard Magnetic DNA Purification System for Food includes PMPs, lysis buffer A and B, RNase A solution, and protocol. DNA Quantitation System detects picogram amounts of linear dsDNA. It includes buffer solution, T4 DNA polymerase, sodium pyrophosphate, NDPK, DNA standard, reagent, and more. SV 96 Total RNA Isolation System prepares RNA from cultured and white blood cells and tissue without phenol-chloroform or ethanol.
Accessories information: MagneSphere magnetic separation stands are available separately.