EasySep™ Human Progenitor Cell Enrichment Kit II
Supplier: STEMCELL Technologies
17936 17936RF
MSPP-17936EA 1429.23 USD
MSPP-17936 MSPP-17936RF
EasySep™ Human Progenitor Cell Enrichment Kit II
Cell Separation Media
Immunomagnetic enrichment of untouched human hematopoietic progenitor cells.
  • Fast, easy-to-use, and column-free procedure
  • Up to 95% purity
  • Untouched, viable cells
The EasySep™ Human Progenitor Cell Enrichment Kit II is designed to isolate hematopoietic progenitor cells from fresh cord blood and other cell preparations by negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').