EasySep™ Human Naïve B Cell Enrichment Kits
Supplier: STEMCELL Technologies
19254 19254RF
MSPP-19254EA 1466.15 USD
MSPP-19254 MSPP-19254RF
EasySep™ Human Naïve B Cell Enrichment Kits
Cell Separation Media
Immunomagnetic isolation of untouched human naïve B cells
  • Fast, easy-to-use and column-free
  • Up to 98% purity
  • Isolated cells are untouched
The EasySep™ Human Naïve B Cell Enrichment Kit is designed to isolate naïve B cells from peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-naïve B cells and dextran-coated magnetic particles. The labeled cells are separated using the EasySep™ magnet without the use of columns. The desired cells are poured off into a new tube.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').