EasySep™ Direct Human Total Lymphocyte Isolation Kits
Supplier: STEMCELL Technologies
19655 19655RF
MSPP-19655EA 864.62 USD
MSPP-19655 MSPP-19655RF
EasySep™ Direct Human Total Lymphocyte Isolation Kits
Cell Separation Media
Immunomagnetic isolation of total lymphocytes directly from whole blood
  • >99.9% RBC depletion without the need for density gradient centrifugation, sedimentation or lysis
  • Fast, easy-to-use and column-free
  • Up to 96% purity of isolated cells
  • Isolated cells are untouched
The EasySep™ Direct Human Total Lymphocyte Isolation Kit isolates highly purified lymphocytes directly from human whole blood by immunomagnetic negative selection. Isolated cells are untouched and available for direct use in flow cytometry, culture​,​ DNA/RNA extraction or other downstream applications.​
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').