Cellulose Acetate Membranes
Supplier: Sartorius
12303-142------G 12303-142------N 12303-293------G 12303-293------N 12342-142------G 12342-293------G 11104-142------G 11104-142------N 11104-293------G 11104-293------N 11105-142------G 11105-142------N 11105-293------G 11106-142------G 11106-142------N 11106-293------G 11106-293------N 11107-142------G 11107-142------N 11107-293------G 11107-293------N
77578-736PK 265.48 USD
77578-736 77578-738 77578-740 77578-742 77578-744 77578-746 77578-672 77578-674 77578-676 77578-678 77578-680 77578-682 77578-684 77578-686 77578-688 77578-690 77578-692 77578-694 77578-696 77578-698 77578-700
Cellulose Acetate Membranes
Filters Membrane Filters
Cellulose acetate membranes, hydrophilic, ideally suited for pressure filtration.
  • High flow rates and thermal stability
  • Excellently suited for pressure filtration devices
  • Compatible with aqueous solutions, oils, alcohols and other organic solvents
  • Discs filter form
They combine thermal stability with exceptionally low adsorption characteristics. They have high flow rates thanks to their symmetrical structure and are compatible with aqueous solutions (pH 4 to 8), oils, alcohols and other organic solvents. Available in different pore and diameter sizes from 0.2 to 5 μm and 13 to 293 mm respectively.