4-(4-Chlorophenyl)-2-methyl-3-butyn-2-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
321621-5G
321621-25G
321621-1G
321621-500MG
77634-752EA
329
USD
77634-752
77658-706
77642-012
77650-058
4-(4-Chlorophenyl)-2-methyl-3-butyn-2-ol ≥95%
4-(4-Chlorophenyl)-2-methyl-3-butyn-2-ol
Caution: For Research Use Only
Formula:
C₁₁H₁₁ClO Storage Temperature: Ambient |
MDL Number:
MFCD06803758 CAS Number: 75373-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...