(4'-Chloro-[1,1'-biphenyl]-4-yl)phenylmethanone ≥95%

Supplier: AOBChem USA

Warning

310276-5G 310276-25G 310276-1G 310276-500MG
77632-626EA 1441 USD
77632-626 77654-904 77641-922 77645-936
(4'-Chloro-[1,1'-biphenyl]-4-yl)phenylmethanone ≥95%
(4'-Chloro-[1,1'-biphenyl]-4-yl)phenylmethanone

Caution: For Research Use Only

Formula: C₁₉H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD35171859
CAS Number: 63242-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR