Methyl 2-fluoro-5-(methylsulfanyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

30983-5G 30983-25G 30983-1G 30983-500MG
77632-622EA 933 USD
77632-622 77654-898 77641-916 77645-934
Methyl 2-fluoro-5-(methylsulfanyl)benzoate ≥97%
Methyl 2-fluoro-5-(methylsulfanyl)benzoate

Caution: For Research Use Only

Formula: C₉H₉FO₂S
Storage Temperature: Ambient
MDL Number: MFCD22489098
CAS Number: 1541278-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR