1-[4-Ethoxy-3-(trifluoromethyl)phenyl]-1-propanone ≥95%
Supplier: AOBChem USA
|
Warning
|
309752-5G
309752-25G
309752-1G
309752-500MG
77628-522EA
1484.23
USD
77628-522
77654-896
77641-914
77645-928
1-[4-Ethoxy-3-(trifluoromethyl)phenyl]-1-propanone ≥95%
1-[4-Ethoxy-3-(trifluoromethyl)phenyl]-1-propanone
Caution: For Research Use Only
Formula:
C₁₂H₁₃F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD28849754 CAS Number: 1806688-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...