1-Phenyl-1-hexyn-3-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
258768-5G
258768-25G
77634-516EA
66
USD
77634-516
77654-484
1-Phenyl-1-hexyn-3-ol ≥97%
1-Phenyl-1-hexyn-3-ol
Caution: For Research Use Only
Formula:
C₁₂H₁₄O Storage Temperature: Ambient |
MDL Number:
MFCD00046687 CAS Number: 1817-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...