1-Butyl-3-methylimidazolium tricyanomethanide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

B5487-25G B5487-5G
76219-630EA 294 USD
76219-630 76219-632
1-Butyl-3-methylimidazolium tricyanomethanide ≥98.0% (by HPLC, total nitrogen)
1-Butyl-3-methylimidazolium tricyanomethanide
Formula: C₁₂H₁₅N₅
MW: 229.29 g/mol
MDL Number: MFCD09038877
CAS Number: 878027-73-7

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR