Benorilate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
Aspirin Acetaminophen ester, 4-Acetamidophenyl 2-Acetoxybenzoate
B4827-25G
B4827-5G
TCB4827-25GEA
112
USD
TCB4827-25G
TCB4827-5G
Benorilate ≥98.0% (by HPLC, total nitrogen)
Benorilate
Formula:
C₁₇H₁₅NO₅ MW: 313.31 g/mol |
MDL Number:
MFCD00864257 CAS Number: 5003-48-5 EINECS: 225-674-5 Merck Index: 14,01045 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 178.0 to 182.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...